Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
482404
Gene Name
THAP domain containing 5
Gene Aliases
-
Chromosome Location
Chr.14: 48738882 - 48880295 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628731.3 XP_005628788.1 2 - 3 271 161
XM_022427579.1 XP_022283287.1 2 - 3 314 161
XM_022427580.1 XP_022283288.1 2 - 3 291 161
XM_022427581.1 XP_022283289.1 2 - 3 306 161
XM_539521.6 XP_539521.2 2 - 3 380 161

View Full Product Details