Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINH1
Entrez Gene ID
485187
Gene Name
serpin family H member 1
Gene Aliases
-
Chromosome Location
Chr.21: 23032761 - 23041649 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165888.1 NP_001159360.1 3 - 4 879 73
XM_005633296.3 XP_005633353.1 4 - 5 947 73
GenBank mRNA FN395288.1 - 2 - 3 719 73

View Full Product Details