Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID5A
Entrez Gene ID
483050
Gene Name
AT-rich interaction domain 5A
Gene Aliases
-
Chromosome Location
Chr.17: 34320791 - 34333660 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630383.3 XP_005630440.1 5 - 6 498 69
XM_022404674.1 XP_022260382.1 6 - 7 1908 69
XM_022404675.1 XP_022260383.1 7 - 8 1108 69
XM_022404676.1 XP_022260384.1 4 - 5 1716 69
XM_022404677.1 XP_022260385.1 4 - 5 474 69
XM_022404678.1 XP_022260386.1 6 - 7 877 69
XM_540165.6 XP_540165.2 6 - 7 666 69

View Full Product Details