Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK4
Entrez Gene ID
484354
Gene Name
kallikrein related peptidase 4
Gene Aliases
-
Chromosome Location
Chr.1: 105950948 - 105955464 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197169.1 NP_001184098.1 2 - 3 221 70
XM_005615765.3 XP_005615822.1 3 - 4 381 70
GenBank mRNA HM135452.1 - 2 - 3 221 70

View Full Product Details