Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB2
Entrez Gene ID
484036
Gene Name
zinc finger and BTB domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 41719251 - 41743007 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541153.4 XP_541153.2 - 316 100

View Full Product Details