Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC3L2
Entrez Gene ID
484450
Gene Name
exocyst complex component 3 like 2
Gene Aliases
-
Chromosome Location
Chr.1: 110265054 - 110277895 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541565.4 XP_541565.2 8 - 9 1077 53

View Full Product Details