Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAPTM5
Entrez Gene ID
487324
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
-
Chromosome Location
Chr.2: 70188382 - 70215361 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617684.3 XP_005617741.1 3 - 4 355 78

View Full Product Details