Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA3
Entrez Gene ID
487032
Gene Name
carbonic anhydrase 3
Gene Aliases
-
Chromosome Location
Chr.29: 31935192 - 31945585 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145172.1 NP_001138644.1 6 - 7 695 113

View Full Product Details