Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP7
Entrez Gene ID
486438
Gene Name
THAP domain containing 7
Gene Aliases
-
Chromosome Location
Chr.26: 30498718 - 30501085 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410553.1 XP_022266261.1 3 - 4 430 103
XM_543564.5 XP_543564.2 3 - 4 334 103

View Full Product Details