Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX11
Entrez Gene ID
486745
Gene Name
DEAD/H-box helicase 11
Gene Aliases
DDX12
Chromosome Location
Chr.27: 42067845 - 42102699 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637325.2 XP_005637382.1 2 - 3 329 74
XM_014108492.2 XP_013963967.1 2 - 3 329 74

View Full Product Details