Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP19
Entrez Gene ID
488433
Gene Name
dual specificity phosphatase 19
Gene Aliases
-
Chromosome Location
Chr.36: 26095436 - 26111700 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_545555.5 XP_545555.2 1 - 2 292 66

View Full Product Details