Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGT6
Entrez Gene ID
606867
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.5: 30540461 - 30543878 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003434611.4 XP_003434659.2 3 - 4 468 135
XM_022418401.1 XP_022274109.1 2 - 3 868 135

View Full Product Details