Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX10
Entrez Gene ID
100856251
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
-
Chromosome Location
Chr.5: 57289530 - 57291864 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620422.2 XP_005620479.1 - 172 115

View Full Product Details