Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
475826
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
-
Chromosome Location
Chr.17: 58713304 - 58718795 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630729.3 XP_005630786.1 3 - 4 516 84
XM_005630730.3 XP_005630787.1 3 - 4 433 84
XM_533034.6 XP_533034.1 3 - 4 516 84

View Full Product Details