Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS16
Entrez Gene ID
609906
Gene Name
mitochondrial ribosomal protein S16
Gene Aliases
-
Chromosome Location
Chr.4: 23827690 - 23829319 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847254.5 XP_852347.3 2 - 3 480 89

View Full Product Details