Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF20
Entrez Gene ID
609780
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
-
Chromosome Location
Chr.25: 40148578 - 40159975 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847118.4 XP_852211.1 2 - 3 266 96

View Full Product Details