Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB31
Entrez Gene ID
608485
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 75678896 - 75807716 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845517.5 XP_850610.2 2 - 3 232 69

View Full Product Details