Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERP29
Entrez Gene ID
477482
Gene Name
endoplasmic reticulum protein 29
Gene Aliases
-
Chromosome Location
Chr.26: 9608352 - 9616711 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410798.1 XP_022266506.1 - 618 86

View Full Product Details