Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
612555
Gene Name
MAGI family member, X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 42267054 - 42269303 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014111385.1 XP_013966860.1 4 - 5 251 90

View Full Product Details