Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDP
Entrez Gene ID
612440
Gene Name
norrin cystine knot growth factor NDP
Gene Aliases
-
Chromosome Location
Chr.X: 37949560 - 37975573 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850168.5 XP_855261.1 2 - 3 723 88

View Full Product Details