Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEANC2
Entrez Gene ID
610613
Gene Name
transcription elongation factor A N-terminal and central domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 54924184 - 54960956 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620262.3 XP_005620319.1 4 - 5 783 85
XM_005620263.3 XP_005620320.1 4 - 5 828 85
XM_005620264.3 XP_005620321.1 4 - 5 831 85
XM_005620265.3 XP_005620322.1 4 - 5 786 85
XM_005620266.3 XP_005620323.1 4 - 5 1006 85
XM_848147.5 XP_853240.1 4 - 5 1003 85

View Full Product Details