Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRADD
Entrez Gene ID
611262
Gene Name
TNFRSF1A associated via death domain
Gene Aliases
-
Chromosome Location
Chr.5: 82200430 - 82211430 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620856.3 XP_005620913.1 3 - 4 383 86
XM_014114114.2 XP_013969589.1 2 - 3 620 86
XM_848905.5 XP_853998.2 2 - 3 253 86

View Full Product Details