Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOB3A
Entrez Gene ID
612205
Gene Name
MOB kinase activator 3A
Gene Aliases
MOBKL2A
Chromosome Location
Chr.20: 56940743 - 56951906 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849941.5 XP_855034.3 2 - 3 677 70

View Full Product Details