Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
611457
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
-
Chromosome Location
Chr.1: 103021519 - 103027460 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849126.5 XP_854219.1 2 - 3 386 100

View Full Product Details