Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBL5
Entrez Gene ID
611310
Gene Name
ubiquitin like 5
Gene Aliases
-
Chromosome Location
Chr.20: 51174741 - 51176401 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848963.5 XP_854056.1 4 - 5 345 103

View Full Product Details