Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS23
Entrez Gene ID
479159
Gene Name
ribosomal protein S23
Gene Aliases
-
Chromosome Location
Chr.3: 25208812 - 25210492 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618155.3 XP_005618212.1 1 - 1 433 88

View Full Product Details