Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FEZF2
Entrez Gene ID
476568
Gene Name
FEZ family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.20: 28704287 - 28708620 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014122400.2 XP_013977875.1 1 - 2 586 80

View Full Product Details