Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIM
Entrez Gene ID
477991
Gene Name
vimentin
Gene Aliases
-
Chromosome Location
Chr.2: 19671316 - 19679367 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287023.1 NP_001273952.1 - 1150 140
GenBank mRNA DN371136.1 - - 454 140

View Full Product Details