Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIP7
Entrez Gene ID
479673
Gene Name
nucleolar pre-rRNA processing protein NIP7
Gene Aliases
-
Chromosome Location
Chr.5: 80287699 - 80316350 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287127.1 NP_001274056.1 2 - 3 176 137
XM_022418020.1 XP_022273728.1 2 - 3 251 137
GenBank mRNA CF410252.1 - 2 - 3 172 138

View Full Product Details