Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASD2
Entrez Gene ID
481284
Gene Name
RASD family member 2
Gene Aliases
-
Chromosome Location
Chr.10: 28626761 - 28637861 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538405.5 XP_538405.3 2 - 3 694 134

View Full Product Details