Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGN
Entrez Gene ID
612577
Gene Name
gametogenetin
Gene Aliases
-
Chromosome Location
Chr.1: 114615817 - 114618950 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014120664.2 XP_013976139.1 1 - 1 484 146

View Full Product Details