Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYRK3
Entrez Gene ID
480010
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases
-
Chromosome Location
Chr.7: 5816839 - 5826194 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003434949.4 XP_003434997.2 - 1377 149
XM_014115059.2 XP_013970534.1 - 1389 149
XM_022420843.1 XP_022276551.1 - 1359 149
XM_022420844.1 XP_022276552.1 - 1275 149

View Full Product Details