Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FCRLB
Entrez Gene ID
609689
Gene Name
Fc receptor like B
Gene Aliases
-
Chromosome Location
Chr.38: 20943351 - 20950836 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022415320.1 XP_022271028.1 3 - 3 242 157

View Full Product Details