Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100856603
Entrez Gene ID
100856603
Gene Name
zinc finger protein 850
Gene Aliases
ZNF2
Chromosome Location
Chr.17: 34847821 - 34900990 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022404359.1 XP_022260067.1 5 - 5 2144 181

View Full Product Details