Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM51
Entrez Gene ID
607267
Gene Name
transmembrane protein 51
Gene Aliases
-
Chromosome Location
Chr.2: 82251395 - 82307008 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617974.3 XP_005618031.1 3 - 3 530 131
XM_005617975.3 XP_005618032.1 3 - 3 594 131
XM_014111400.2 XP_013966875.1 3 - 3 462 131
XM_022414389.1 XP_022270097.1 3 - 3 558 131
XM_022414391.1 XP_022270099.1 4 - 4 668 131
XM_022414394.1 XP_022270102.1 4 - 4 618 131
XM_843883.5 XP_848976.1 3 - 3 544 131

View Full Product Details