Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBTD2
Entrez Gene ID
489120
Gene Name
ubiquitin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.4: 39992560 - 40064512 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417694.1 XP_022273402.1 3 - 3 408 91
XM_022417695.1 XP_022273403.1 4 - 4 536 91
XM_022417696.1 XP_022273404.1 3 - 3 489 91
XM_546238.5 XP_546238.2 3 - 3 521 91

View Full Product Details