Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM86B
Entrez Gene ID
611345
Gene Name
transmembrane protein 86B
Gene Aliases
-
Chromosome Location
Chr.1: 102427967 - 102430426 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616177.3 XP_005616234.1 3 - 3 1236 143
XM_005616178.3 XP_005616235.1 3 - 3 1233 143

View Full Product Details