Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIO3
Entrez Gene ID
612596
Gene Name
iodothyronine deiodinase 3
Gene Aliases
-
Chromosome Location
Chr.8: 69696779 - 69698880 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164188.1 NP_001157660.1 1 - 1 668 96

View Full Product Details