Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
APOE
Entrez Gene ID
476438
Gene Name
apolipoprotein E
Gene Aliases
Apo-E
Chromosome Location
Chr.1: 110525717 - 110528534 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616460.3 XP_005616517.2 3 - 3 360 188
XM_022425020.1 XP_022280728.1 3 - 3 359 188
XM_533644.6 XP_533644.1 3 - 3 373 188
XM_860950.4 XP_866043.1 3 - 3 387 188

View Full Product Details