Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
OPRD1
Entrez Gene ID
487329
Gene Name
opioid receptor delta 1
Gene Aliases
-
Chromosome Location
Chr.2: 71685129 - 71721692 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544455.6 XP_544455.3 3 - 3 675 147

View Full Product Details