Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM64
Entrez Gene ID
487040
Gene Name
transmembrane protein 64
Gene Aliases
-
Chromosome Location
Chr.29: 36005600 - 36030366 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014109313.2 XP_013964788.1 3 - 3 565 137

View Full Product Details