Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA30
Entrez Gene ID
612880
Gene Name
N-alpha-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
NAT12
Chromosome Location
Chr.8: 32939442 - 32959854 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846834.5 XP_851927.1 4 - 5 1217 155

View Full Product Details