Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAG1
Entrez Gene ID
486960
Gene Name
PLAG1 zinc finger
Gene Aliases
-
Chromosome Location
Chr.29: 7563804 - 7608819 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003433656.4 XP_003433704.1 4 - 4 1269 94
XM_005638000.3 XP_005638057.1 5 - 5 1374 94
XM_022412467.1 XP_022268175.1 3 - 3 910 94

View Full Product Details