Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX1
Entrez Gene ID
482587
Gene Name
ALX homeobox 1
Gene Aliases
CART1
Chromosome Location
Chr.15: 26838615 - 26858786 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022403248.1 XP_022258956.1 2 - 3 1871 129

View Full Product Details