Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
100329125
Gene Name
kallikrein related peptidase 9
Gene Aliases
-
Chromosome Location
Chr.1: 105868147 - 105873147 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171773.2 NP_001165244.2 3 - 3 440 130
GenBank mRNA GQ303574.1 - 3 - 3 440 130
GQ303577.1 - 2 - 2 617 130

View Full Product Details