Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB43
Entrez Gene ID
612870
Gene Name
zinc finger and BTB domain containing 43
Gene Aliases
-
Chromosome Location
Chr.9: 56437649 - 56464617 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005625348.3 XP_005625405.1 3 - 3 1530 121
XM_005625349.3 XP_005625406.1 2 - 2 1418 121
XM_022423809.1 XP_022279517.1 3 - 3 2044 121
XM_022423810.1 XP_022279518.1 3 - 3 1556 121
XM_846547.5 XP_851640.2 3 - 3 1543 121

View Full Product Details