Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
486658
Gene Name
RERG like
Gene Aliases
-
Chromosome Location
Chr.27: 28943933 - 28959374 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543785.6 XP_543785.4 4 - 5 614 186

View Full Product Details