Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB1B
Entrez Gene ID
610762
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.18: 51026975 - 51034356 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022405629.1 XP_022261337.1 5 - 6 600 136
XM_847452.5 XP_852545.1 5 - 6 538 136

View Full Product Details