Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SALL3
Entrez Gene ID
607126
Gene Name
spalt like transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.1: 1413991 - 1421516 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426699.1 XP_022282407.1 - 3306 180

View Full Product Details