Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM102
Entrez Gene ID
111089959
Gene Name
transmembrane protein 102
Gene Aliases
-
Chromosome Location
Chr.5: 32365331 - 32367785 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022418517.1 XP_022274225.1 2 - 2 1971 124
XM_022418518.1 XP_022274226.1 2 - 2 1806 124

View Full Product Details